4-Phenylbenzonitrile

Supplier: Matrix Scientific
Warning

Synonyms: 4-Biphenylcarbonitrile, 4-Cyanobiphenyl, p-Phenylbenzonitrile, [1,1'-Biphenyl]-4-carbonitrile

005696-5G 005696-25G
101789-374EA 29.23 USD
101789-374 101789-376
4-Phenylbenzonitrile
4-Phenylbenzonitrile

Hazard code(s): Irritant

Formula: C₁₃H₉N
MW: 179.22 g/mol
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00001821
CAS Number: 2920-38-9
EINECS: 220-860-2
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 85-87°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR