3-Methoxyphenethylamine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(3-Methoxyphenyl)ethylamine

005691-5G 005691-25G 005691-100G
101789-352EA 41.54 USD
101789-352 101789-354 101789-356
3-Methoxyphenethylamine ≥97%
3-Methoxyphenethylamine

Hazard code(s): Irritant

Formula: C₉H₁₃NO
MW: 151.21 g/mol
Boiling Pt: 118…119 °C (6mm)
Density: 1.04 g/cm³
MDL Number: MFCD00008187
CAS Number: 2039-67-0
EINECS: 218-017-9

Order Now

Specification Test Results

Boiling point 118-119°C/6
Density 1.04

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR