1-(4-Methoxyphenyl)imidazole ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(4-Methoxyphenyl)-1H-imidazole

005667-1G 005667-5G
101789-314EA 52.31 USD
101789-314 101789-316
1-(4-Methoxyphenyl)imidazole ≥98%
1-(4-Methoxyphenyl)imidazole

Hazard code(s): Irritant

Formula: C₁₀H₁₀N₂O
MW: 174.2 g/mol
Boiling Pt: 120 °C (0.1mm)
Melting Pt: 64…66 °C
MDL Number: MFCD00060492
CAS Number: 10040-95-6

Order Now

Specification Test Results

Boiling point 120°C/0.1mm
Melting point 64-66°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR