1,4-Diiodotetrafluorobenzene ≥98%
Supplier: Matrix Scientific
|
Warning
|
005637-1G
005637-5G
101789-272EA
44.62
USD
101789-272
101789-274
1,4-Diiodotetrafluorobenzene ≥98%
1,4-Diiodotetrafluorobenzene
Hazard code(s): Irritant, Refrigerate
Formula:
C₆F₄I₂ Melting Pt: 107…108 °C |
MDL Number:
MFCD00001055 CAS Number: 392-57-4 EINECS: 206-877-8 |
Specification Test Results
Melting point | 107-108°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...