1,2-Dichloro-1,1,2-trifluoro-2-(trifluoromethoxy)ethane ≥98%

Supplier: Matrix Scientific

Synonyms: 1,2-Dichloro-1,2,2-trifluoroethyl trifluoromethyl ether

005632-1G 005632-5G 005632-25G
101789-262EA 38.46 USD
101789-262 101789-264 101789-266
1,2-Dichloro-1,1,2-trifluoro-2-(trifluoromethoxy)ethane ≥98%
1,2-Dichloro-1,1,2-trifluoro-2-(trifluoromethoxy)ethane

Hazard code(s): Irritant

Formula: ClF₂CCClFOCF₃
MW: 236.93 g/mol
MDL Number: MFCD00464450
CAS Number: 2356-53-8
EINECS: 219-094-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR