2,3-Dichlorothiophenol ≥98%

Supplier: Matrix Scientific

Synonyms: 2,3-Dichlorobenzenethiol, 1,2-Dichloro-3-mercaptobenzene, 2,3-Dichlorobenzene-1-thiol

005626-5G 005626-25G
101789-258EA 26.15 USD
101789-258 101789-260
2,3-Dichlorothiophenol ≥98%
2,3-Dichlorothiophenol

Hazard code(s): Irritant, Stench

Formula: C₆H₄Cl₂S
MW: 179.07 g/mol
Boiling Pt: 95…98 °C (0.5mm)
Melting Pt: 55…59 °C
MDL Number: MFCD00041420
CAS Number: 17231-95-7

Order Now

Specification Test Results

Boiling point 95-98°C/0.5
Melting point 55-59°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR