3,4-Dichlorothiophenol ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 1,2-Dichloro-4-mercaptobenzene, 1,2-Dichloro-4-benzenethiol, 3,4-Dichlorobenzenethiol, 3,4-Dichlorobenzene-1-thiol

005623-25G 005623-100G
101789-250EA 176.92 USD
101789-250 101789-252
3,4-Dichlorothiophenol ≥98%
3,4-Dichlorothiophenol

Hazard code(s): Irritant, Stench

Formula: C₆H₄Cl₂S
MW: 179.07 g/mol
Boiling Pt: 76 - 78 °C (0.3 -rr)
Melting Pt: 87 - 89 °C
Density: 1.4175 g/cm³ (20 °C)
MDL Number: MFCD00004840
CAS Number: 5858-17-3
EINECS: 227-485-3

Order Now

Specification Test Results

Boiling point 119-120°C/1
Density 1.407

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR