2,4-Dichlorothiophenol ≥97%

Supplier: Matrix Scientific
Warning

005619-25G 005619-100G
101789-242EA 300 USD
101789-242 101789-244
2,4-Dichlorothiophenol ≥97%
2,4-Dichlorothiophenol

Hazard code(s): Irritant, Stench

Formula: C₆H₄Cl₂S
Boiling Pt: 115 °C (8 mmHg)
Melting Pt: 22 °C
Density: 1.44 g/cm³ (20 °C)
MDL Number: MFCD00041421
CAS Number: 1122-41-4
EINECS: 214-346-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR