3,4-Dichlorophenylhydrazinium chloride ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 3,4-Dichlorophenylhydrazine hydrochloride, 1-(3,4-Dichlorophenyl)hydrazine hydrochloride

005599-5G 005599-25G
101789-224EA 38.46 USD
101789-224 101789-226
3,4-Dichlorophenylhydrazinium chloride ≥97% for synthesis
3,4-Dichlorophenylhydrazinium chloride

Hazard code(s): Irritant

Formula: C₆H₇Cl₃N₂
Melting Pt: 230 °C (dec.)
MDL Number: MFCD00012937
CAS Number: 19763-90-7
EINECS: 243-278-0

Order Now

Specification Test Results

Melting point 230°C(dec)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR