2,4-Difluorophenylhydrazinium chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2,4-Difluorophenylhydrazine hydrochloride, 1-(2,4-Difluorophenyl)hydrazine hydrochloride

005597-5G 005597-25G 005597-100G
101789-218EA 15.38 USD
101789-218 101789-220 101789-222
2,4-Difluorophenylhydrazinium chloride ≥97%
2,4-Difluorophenylhydrazinium chloride

Hazard code(s): Irritant

Formula: C₆H₇ClF₂N₂
MW: 180.58 g/mol
Melting Pt: 250…260 °C
MDL Number: MFCD00013383
CAS Number: 51523-79-6

Order Now

Specification Test Results

Melting point 250-260°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR