2,5-Difluorophenylhydrazine ≥98%

Supplier: Matrix Scientific

005596-5G 005596-25G
101789-216EA 64.62 USD
101789-216 102832-772
2,5-Difluorophenylhydrazine ≥98%
2,5-Difluorophenylhydrazine

Hazard code(s): Irritant

Formula: C₆H₆F₂N₂
MW: 144.12 g/mol
MDL Number: MFCD00013384
CAS Number: 97108-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR