3,4-Difluorophenylboronic acid ≥98%

Supplier: Matrix Scientific

005592-25G 005592-100G
101789-206EA 89.23 USD
101789-206 101789-208
3,4-Difluorophenylboronic acid ≥98%
3,4-Difluorophenylboronic acid

Hazard code(s): Store in Freezer

Formula: C₆H₅BF₂O₂
Melting Pt: 305…310 °C
Storage Temperature: Ambient
MDL Number: MFCD00807405
CAS Number: 168267-41-2

Order Now

Specification Test Results

Melting point 305-310°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR