2,6-Difluorophenylboronic acid ≥98% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: Dihydroxy(2,6-difluorophenyl)borane, 2,6-Difluorobenzeneboronic acid, (2,6-Difluorophenyl)metaboric acid

005590-5G 005590-25G
101789-196EA 55.38 USD
101789-196 101789-198
2,6-Difluorophenylboronic acid ≥98% for synthesis
2,6-Difluorophenylboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₆H₅BF₂O₂
Melting Pt: 144…145 °C
MDL Number: MFCD00792436
CAS Number: 162101-25-9

Order Now

Specification Test Results

Melting point 144-145°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR