(3,4-Difluorophenyl)acetic acid ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 3,4-Difluorophenylacetic acid

005581-5G 005581-25G 005581-100G
101789-182EA 21.54 USD
101789-182 101789-184 101789-186
(3,4-Difluorophenyl)acetic acid ≥98%
(3,4-Difluorophenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₃F₂)CH₂COOH
MW: 172.13 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 46…50 °C
MDL Number: MFCD00010002
CAS Number: 658-93-5
EINECS: 211-527-2

Order Now

Specification Test Results

Melting point 46-50°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR