(2,6-Difluorophenyl)acetic acid ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 2,6-Difluorophenylacetic acid

005580-5G 005580-25G
101789-178EA 21.54 USD
101789-178 101789-180
(2,6-Difluorophenyl)acetic acid ≥97%
(2,6-Difluorophenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₃F₂)CH₂COOH
MW: 172.13 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 100 - 102 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010001
CAS Number: 85068-28-6
EINECS: 285-289-3

Order Now

Specification Test Results

Melting point 100-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR