(2,5-Difluorophenyl)acetic acid ≥95%

Supplier: Matrix Scientific
Warning

Synonyms: 2,5-Difluorophenylacetic acid

005579-25G 005579-100G
101789-174EA 192.31 USD
101789-174 101789-176
(2,5-Difluorophenyl)acetic acid ≥95%
(2,5-Difluorophenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₃F₂)CH₂COOH
MW: 172.13 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 126…128 °C
MDL Number: MFCD00010000
CAS Number: 85068-27-5
EINECS: 285-288-8

Order Now

Specification Test Results

Melting point 126-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR