(2,4-Difluorophenyl)acetic acid ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 2,4-Difluorophenylacetic acid

005578-25G 005578-100G
101789-168EA 75.38 USD
101789-168 101789-170
(2,4-Difluorophenyl)acetic acid ≥97%
(2,4-Difluorophenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₃F₂)CH₂COOH
MW: 172.13 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 117 - 119 °C
MDL Number: MFCD00009999
CAS Number: 81228-09-3
EINECS: 279-709-4

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR