(2,4-Dichlorophenyl)acetic acid ≥99%

Supplier: Matrix Scientific
Warning

Synonyms: DCAA, 2,4-Dichlorophenylacetic acid

005574-25G 005574-100G
101789-154EA 29.23 USD
101789-154 101789-156
(2,4-Dichlorophenyl)acetic acid ≥99%
(2,4-Dichlorophenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₃Cl₂)CH₂COOH
MW: 205.04 g/mol
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 131…133 °C
MDL Number: MFCD00004318
CAS Number: 19719-28-9
EINECS: 243-248-7

Order Now

Specification Test Results

Melting point 131-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR