Bis(4-fluorophenyl) sulfone ≥98%
Supplier: Matrix Scientific
Synonyms:
4-Fluorophenyl sulphone, 4,4'-Difluoro diphenyl sulphone, 1-Fluor-4-((4-fluorphenyl)sulphonyl)benzol, 4,4'-sulphonylbis(fluorobenzene)
005568-25G
005568-100G
101789-142EA
44.62
USD
101789-142
101789-144
Bis(4-fluorophenyl) sulfone ≥98%
Bis(4-fluorophenyl)sulphone
Hazard code(s): Irritant
Formula:
C₁₂H₈F₂O₂S MW: 254.26 g/mol Melting Pt: 98…99 °C |
MDL Number:
MFCD00000350 CAS Number: 383-29-9 EINECS: 206-847-4 |
Specification Test Results
Melting point | 98-99°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...