3,4-Difluorophenol ≥98%

Supplier: Matrix Scientific
Warning

005565-25G 005565-100G
101789-138EA 55.38 USD
101789-138 101789-140
3,4-Difluorophenol ≥98%
3,4-Difluorophenol

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 32 - 36 °C
MDL Number: MFCD00010315
CAS Number: 2713-33-9
UN: 2921
ADR: 8,II

Order Now

Specification Test Results

Boiling point 85°C/20mm
Melting point 34-38°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR