2,3-Difluorophenol ≥98%

Supplier: Matrix Scientific

005563-5G 005563-25G 005563-100G
101789-132EA 290.77 USD
101789-132 101789-134 101789-136
2,3-Difluorophenol ≥98%
2,3-Difluorophenol

Hazard code(s): Irritant

Formula: C₆H₄F₂O
Boiling Pt: 54 °C (25mm)
Melting Pt: 39…42 °C
MDL Number: MFCD00010262
CAS Number: 6418-38-8
UN: 2921
ADR: 8,II

Order Now

Specification Test Results

Boiling point 54°C/25mm
Melting point 39-42°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR