Furan-2-boronic acid tech, tech.
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
Furan-2-ylboronic acid
005543-5G
005543-25G
101789-102EA
23.08
USD
101789-102
101789-104
Furan-2-boronic acid tech, tech.
Furan-2-boronic acid
Store under ARGON
Hazard code(s): Irritant, Store in Freezer, Air Sensitive, Stored under Argon
Formula:
C₄H₅BO₃ MW: 111.89 g/mol Melting Pt: 120…121 °C |
MDL Number:
MFCD00799544 CAS Number: 13331-23-2 |
Specification Test Results
Melting point | 120-121°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...