4-((Trifluoromethyl)thio)benzoic acid ≥97%

Supplier: Matrix Scientific

005522-25G 005522-100G
101789-086EA 83.08 USD
101789-086 101789-088
4-((Trifluoromethyl)thio)benzoic acid ≥97%
4-((Trifluoromethyl)thio)benzoic acid

Hazard code(s): Irritant

Formula: C₈H₅F₃O₂S
MW: 222.19 g/mol
MDL Number: MFCD00040906
CAS Number: 330-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR