2-(Trifluoromethyl)benzyl alcohol ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(Hydroxymethyl)benzotrifluoride, (2-(Trifluoromethyl)phenyl)methanol

005510-25G 005510-100G
101789-076EA 36.92 USD
101789-076 101789-078
2-(Trifluoromethyl)benzyl alcohol ≥97%
2-(Trifluoromethyl)benzyl alcohol

Hazard code(s): Irritant

Formula: C₈H₇F₃O
MW: 176.14 g/mol
Boiling Pt: 90 °C (20mm)
Melting Pt: 4 °C
Density: 1.32 g/cm³ (20 °C)
MDL Number: MFCD00004621
CAS Number: 346-06-5
EINECS: 206-467-9

Order Now

Specification Test Results

Boiling point 90°C/20mm
Density 1.326
ND20 1.468

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR