4-(Trifluoromethyl)benzyl alcohol ≥98% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: (4-(Trifluoromethyl)phenyl)methanol, 4-(Hydroxymethyl)benzotrifluoride

005500-5G 005500-25G 005500-100G
101789-066EA 24.62 USD
101789-066 101789-068 101789-070
4-(Trifluoromethyl)benzyl alcohol ≥98% for synthesis
4-(Trifluoromethyl)benzyl alcohol

Hazard code(s): Irritant, Store in Freezer

Formula: C₈H₇F₃O
Boiling Pt: 214 °C (1013 hPa)
Melting Pt: 23 °C
Density: 1.28 g/cm³ (20 °C)
MDL Number: MFCD00004661
CAS Number: 349-95-1
EINECS: 206-494-6

Order Now

Specification Test Results

Boiling point 78-80°C/4mm
Density 1.286
ND20 1.459


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR