α'-Bromo-α,α,α-trifluoro-m-xylene ≥98%

Supplier: Matrix Scientific
Danger

005493-25G 005493-100G
101789-050EA 41.54 USD
101789-050 101789-052
α'-Bromo-α,α,α-trifluoro-m-xylene ≥98%
α'-Bromo-α,α,α-trifluoro-m-xylene

Hazard code(s): Corrosive, Lachrymator

Formula: C₈H₆BrF₃
Boiling Pt: 50 °C (0.3 -rr)
Density: 1.56 g/cm³ (20 °C)
MDL Number: MFCD00000395
CAS Number: 402-23-3
EINECS: 206-938-9
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 69°C/4mm
Density 1.565
ND20 1.492


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR