3-(Trifluoromethyl)benzyl alcohol ≥97%

Supplier: Matrix Scientific

005492-25G 005492-100G
101789-046EA 41.54 USD
101789-046 101789-048
3-(Trifluoromethyl)benzyl alcohol ≥97%
3-(Trifluoromethyl)benzyl alcohol

Hazard code(s): Irritant

Formula: C₈H₇F₃O
Boiling Pt: 68 °C (1013 hPa)
Density: 1.2949 g/cm³ (21 °C)
MDL Number: MFCD00004645
CAS Number: 349-75-7
EINECS: 206-489-9

Order Now

Specification Test Results

Boiling point 68°C/2mm
Density 1.295
ND20 1.46


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR