4-(Trifluoromethyl)thiophenol ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Mercaptobenzotrifluoride, 4-(Trifluoromethyl)benzene-1-thiol, 4-(Trifluoromethyl)benzenethiol
005488-1G
005488-25G
101789-036EA
64.62
USD
101789-036
101789-040
4-(Trifluoromethyl)thiophenol ≥95%
4-(Trifluoromethyl)thiophenol
Hazard code(s): Irritant-Harmful, Stench
Formula:
C₇H₅F₃S MW: 178.18 g/mol Boiling Pt: 66…68 °C (15mm) Density: 1.31 g/cm³ (25 °C) |
MDL Number:
MFCD00042320 CAS Number: 825-83-2 UN: 2810 ADR: 6.1,III |
Specification Test Results
Boiling point | 66-68°C/15m |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...