2-(Trifluoromethyl)thiophenol ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 2-(Trifluoromethyl)benzene-1-thiol, 2-(Trifluoromethyl)benzenethiol, 2-Mercaptobenzotrifluoride

005479-1G 005479-5G
101789-026EA 26.15 USD
101789-026 101789-028
2-(Trifluoromethyl)thiophenol ≥97%
2-(Trifluoromethyl)thiophenol

Hazard code(s): Irritant, Stench

Formula: C₇H₅F₃S
MW: 178.18 g/mol
Boiling Pt: 50…52 °C (15mm)
MDL Number: MFCD00068896
CAS Number: 13333-97-6
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 50-52°C/15m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR