α,α,α-Trifluoro-o-toluic acid ≥98%
Supplier: Matrix Scientific
|
Warning
|
005477-25G
005477-100G
101789-020EA
27.69
USD
101789-020
101789-022
α,α,α-Trifluoro-o-toluic acid ≥98%
α,α,α-Trifluoro-o-toluic acid
Hazard code(s): Irritant
Formula:
C₈H₅F₃O₂ Boiling Pt: 247 °C (753mm) Melting Pt: 109…113 °C |
MDL Number:
MFCD00002476 CAS Number: 433-97-6 EINECS: 207-093-9 |
Specification Test Results
Boiling point | 247°C/753mm |
Melting point | 109-113°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...