α,α,α-Trifluoro-m-toluic acid ≥99%

Supplier: Matrix Scientific
Warning

005476-25G 005476-100G
101789-016EA 21.54 USD
101789-016 101789-018
α,α,α-Trifluoro-m-toluic acid ≥99%
α,α,α-Trifluoro-m-toluic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₈H₅F₃O₂
Boiling Pt: 238.5 °C (775mm)
Melting Pt: 105…106 °C
Density: 1.371 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002519
CAS Number: 454-92-2
EINECS: 207-230-2

Order Now

Specification Test Results

Boiling point 238.5°C/775
Melting point 105-106°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR