α,α,α-Trifluoro-o-toluonitrile ≥98%
Supplier: Matrix Scientific
005473-25G
005473-100G
101789-012EA
30.77
USD
101789-012
101789-014
α,α,α-Trifluoro-o-toluonitrile ≥98%
α,α,α-Trifluoro-o-toluonitrile
Hazard code(s): Toxic, Lachrymator
Formula:
C₈H₄F₃N Boiling Pt: 204 °C (1013 hPa) Melting Pt: 18 °C Density: 1.2886 g/cm³ (20 °C) |
MDL Number:
MFCD00001791 CAS Number: 447-60-9 EINECS: 207-184-3 UN: 3276 ADR: 6.1,III |
Specification Test Results
Boiling point | 204-206°C |
Melting point | 7.5°C |
Density | 1.294 |
ND20 | 1.4632 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...