3-(Trifluoromethyl)phenylhydrazinium chloride ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Trifluoromethyl)phenylhydrazine hydrochloride, 3-Hydrazinobenzotrifluoride hydrochloride, (3-(Trifluoromethyl)phenyl)hydrazine hydrochloride, 1-(3-(Trifluoromethyl)phenyl)hydrazine hydrochloride

005466-1G 005466-5G 005466-25G
101788-998EA 15.38 USD
101788-998 101789-000 101789-002
3-(Trifluoromethyl)phenylhydrazinium chloride ≥95%
3-(Trifluoromethyl)phenylhydrazinium chloride

Hazard code(s): Irritant

Formula: C₇H₈ClF₃N₂
MW: 212.6 g/mol
MDL Number: MFCD00100503
CAS Number: 3107-33-3

Order Now

Specification Test Results

Boiling point 140°C/30mm
Density 1.348
ND20 1.504

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR