4-(Trifluoromethyl)phenylhydrazine ≥97%
Supplier: Matrix Scientific
Synonyms:
1-(4-(Trifluoromethyl)phenyl)hydrazine, 4-Hydrazinobenzotrifluoride
005459-5G
005459-25G
005459-100G
101788-982EA
150.77
USD
101788-982
101788-984
101788-986
4-(Trifluoromethyl)phenylhydrazine ≥97%
4-(Trifluoromethyl)phenylhydrazine
Hazard code(s): Irritant-Harmful, Store in Freezer
Formula:
C₇H₇F₃N₂ MW: 176.14 g/mol Boiling Pt: 118…122 °C (17mm) Melting Pt: 63…65 °C |
MDL Number:
MFCD00042508 CAS Number: 368-90-1 |
Specification Test Results
Boiling point | 118-122°C/1 |
Melting point | 63-65°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...