2-(Trifluoromethyl)phenylboronic acid ≥98%

Supplier: Matrix Scientific

Synonyms: (2-Trifluoromethyl)phenylboronic acid, (2-(Trifluoromethyl)phenyl)metaboric acid, Dihydroxy(2-(trifluoromethyl)phenyl)borane, 2-(Trifluoromethyl)benzeneboronic acid

005444-25G 005444-100G
101788-962EA 52.31 USD
101788-962 101788-964
2-(Trifluoromethyl)phenylboronic acid ≥98%
2-(Trifluoromethyl)phenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₇H₆BF₃O₂
MW: 189.93 g/mol
Melting Pt: 108…110 °C
MDL Number: MFCD00236059
CAS Number: 1423-27-4

Order Now

Specification Test Results

Melting point 108-110°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR