6-(Trifluoromethyl)indole ≥97%

Supplier: Matrix Scientific

005439-250MG 005439-1G
101788-954EA 44.62 USD
101788-954 101788-956
6-(Trifluoromethyl)indole ≥97%
6-(Trifluoromethyl)indole

Hazard code(s): Irritant, Store in Freezer

Formula: C₉H₆F₃N
MW: 185.15 g/mol
Melting Pt: 101…105 °C
MDL Number: MFCD00272316
CAS Number: 13544-43-9

Order Now

Specification Test Results

Melting point 101-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR