4-(Trifluoromethyl)phenylboronic acid ≥97%

Supplier: Matrix Scientific

005437-25G 005437-100G
101788-950EA 101.54 USD
101788-950 101788-952
4-(Trifluoromethyl)phenylboronic acid ≥97%
4-(Trifluoromethyl)phenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₇H₆BF₃O₂
Melting Pt: 245…250 °C
MDL Number: MFCD00151855
CAS Number: 128796-39-4

Order Now

Specification Test Results

Melting point 245-250°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR