2-(Trifluoromethyl)phenylhydrazine ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(2-(Trifluoromethyl)phenyl)hydrazine, 2-Hydrazinobenzotrifluoride

005406-1G 005406-5G
101788-874EA 46.15 USD
101788-874 101788-876
2-(Trifluoromethyl)phenylhydrazine ≥98%
2-(Trifluoromethyl)phenylhydrazine

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₇H₇F₃N₂
MW: 176.14 g/mol
Melting Pt: 55…58 °C
MDL Number: MFCD00052687
CAS Number: 365-34-4

Order Now

Specification Test Results

Melting point 55-58°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR