(±)-4-(Trifluoromethyl)mandelic acid ≥98%

Supplier: Matrix Scientific

Synonyms: (±)-4-(Trifluoromethyl)phenylglycolic acid, 2-Hydroxy-2-(4-(trifluoromethyl)phenyl)acetic acid

005402-1G 005402-5G 005402-25G
101788-860EA 13.85 USD
101788-860 101788-862 101788-864
(±)-4-(Trifluoromethyl)mandelic acid ≥98%
(±)-4-(Trifluoromethyl)mandelic acid

Hazard code(s): Irritant

Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 132…133 °C
MDL Number: MFCD00009724
CAS Number: 395-35-7
EINECS: 206-900-1

Order Now

Specification Test Results

Melting point 132-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR