4-(Trifluoromethyl)benzophenone ≥97%

Supplier: Matrix Scientific

Synonyms: Phenyl(4-(trifluoromethyl)phenyl)methanone

005393-5G 005393-25G
101788-838EA 55.38 USD
101788-838 101788-840
4-(Trifluoromethyl)benzophenone ≥97%
4-(Trifluoromethyl)benzophenone

Hazard code(s): Irritant

Formula: C₁₄H₉F₃O
MW: 250.22 g/mol
Boiling Pt: 170 °C (21 mmHg)
Melting Pt: 114…115 °C
MDL Number: MFCD00000400
CAS Number: 728-86-9
EINECS: 211-974-3

Order Now

Specification Test Results

Melting point 114-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR