3-(Trifluoromethyl)benzophenone ≥98%

Supplier: Matrix Scientific

005392-5G 005392-25G
101788-834EA 44.62 USD
101788-834 101788-836
3-(Trifluoromethyl)benzophenone ≥98%
3-(Trifluoromethyl)benzophenone

Hazard code(s): Irritant

Formula: C₁₄H₉F₃O
MW: 250.22 g/mol
Boiling Pt: 160…162 °C (14 mmHg)
Melting Pt: 51…52 °C
MDL Number: MFCD00000389
CAS Number: 728-81-4
EINECS: 211-973-8

Order Now

Specification Test Results

Melting point 51-52°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR