2-(Trifluoromethyl)benzophenone ≥98%

Supplier: Matrix Scientific

005391-5G 005391-25G
101788-830EA 75.38 USD
101788-830 101788-832
2-(Trifluoromethyl)benzophenone ≥98%
2-(Trifluoromethyl)benzophenone

Hazard code(s): Irritant

Formula: C₁₄H₉F₃O
Boiling Pt: 175 - 178 °C (31 -rr)
Melting Pt: 62 °C
MDL Number: MFCD00000377
CAS Number: 727-99-1
EINECS: 211-972-2

Order Now

Specification Test Results

Melting point 57-59°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR