4-(Trifluoromethyl)benzamide ≥99%

Supplier: Matrix Scientific

005390-5G 005390-25G
101788-826EA 21.54 USD
101788-826 101788-828
4-(Trifluoromethyl)benzamide ≥99%
4-(Trifluoromethyl)benzamide

Hazard code(s): Irritant

Formula: C₈H₆F₃NO
MW: 189.14 g/mol
Melting Pt: 250 - 251 °C
MDL Number: MFCD00007998
CAS Number: 1891-90-3
EINECS: 217-571-9

Order Now

Specification Test Results

Melting point 184-185°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR