2-(Trifluoromethyl)benzamide ≥98%

Supplier: Matrix Scientific

005388-5G 005388-25G
101788-818EA 18.46 USD
101788-818 101788-820
2-(Trifluoromethyl)benzamide ≥98%
2-(Trifluoromethyl)benzamide

Hazard code(s): Irritant

Formula: C₈H₆F₃NO
MW: 189.14 g/mol
Melting Pt: 161…162 °C
MDL Number: MFCD00014798
CAS Number: 360-64-5
EINECS: 206-637-2

Order Now

Specification Test Results

Melting point 161-162°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR