5-(Trifluoromethyl)uracil ≥97%

Supplier: Matrix Scientific

Synonyms: Trifluorothymine, 5-(Trifluoromethyl)pyrimidine-2,4(1H,3H)-dione

005384-1G 005384-5G 005384-25G
101788-810EA 23.08 USD
101788-810 101788-812 101788-814
5-(Trifluoromethyl)uracil ≥97%
5-(Trifluoromethyl)uracil

Hazard code(s): Irritant

Formula: C₅H₃F₃N₂O₂
MW: 180.09 g/mol
Boiling Pt: 350 °C (1013 hPa)
Melting Pt: 251 - 252 °C
MDL Number: MFCD00006024
CAS Number: 54-20-6
EINECS: 200-197-5

Order Now

Specification Test Results

Melting point 251-254°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR