4'-(Trifluoromethyl)acetanilide ≥98%

Supplier: Matrix Scientific

Synonyms: N-(4-(Trifluoromethyl)phenyl)acetamide, 4'-(Trifluoromethyl)acetanilide, 4-Acetamidobenzotrifluoride, alpha,alpha,alpha-Trifluoro-p-acetotoluidide

005383-5G 005383-25G
101788-806EA 29.23 USD
101788-806 101788-808
4'-(Trifluoromethyl)acetanilide ≥98%
4'-(Trifluoromethyl)acetanilide

Hazard code(s): Irritant

Formula: C₉H₈F₃NO
MW: 203.16 g/mol
Melting Pt: 208 - 211 °C
MDL Number: MFCD00013562
CAS Number: 349-97-3
EINECS: 206-495-1

Order Now

Specification Test Results

Melting point 152-153°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR