(α,α,α-Trifluoro-4-tolyl)acetic acid ≥97%

Supplier: Matrix Scientific
Warning

005381-5G 005381-25G
101788-796EA 60 USD
101788-796 101788-798
(α,α,α-Trifluoro-4-tolyl)acetic acid ≥97%
(α,α,α-Trifluoro-4-tolyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₄(CF₃))CH₂COOH
Melting Pt: 83…85 °C
MDL Number: MFCD00004352
CAS Number: 32857-62-8
EINECS: 251-263-5

Order Now

Specification Test Results

Melting point 83-85°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR