(α,α,α-Trifluoro-2-tolyl)acetic acid ≥98%

Supplier: Matrix Scientific
Warning

005379-5G 005379-25G
101788-788EA 23.08 USD
101788-788 101788-790
(α,α,α-Trifluoro-2-tolyl)acetic acid ≥98%
(α,α,α-Trifluoro-2-tolyl)acetic acid

Hazard code(s): Irritant

Formula: F₃CC₆H₄CH₂COOH
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 100…102 °C
MDL Number: MFCD00004327
CAS Number: 3038-48-0
EINECS: 221-240-4

Order Now

Specification Test Results

Melting point 100-102°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR