4-(Trifluoromethoxy)benzaldehyde ≥97%

Supplier: Matrix Scientific
Warning

005376-25G 005376-100G
101788-780EA 100 USD
101788-780 101788-782
4-(Trifluoromethoxy)benzaldehyde ≥97%
4-(Trifluoromethoxy)benzaldehyde

Hazard code(s): Irritant

Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Boiling Pt: 93 °C (27mm)
Density: 1.331 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041530
CAS Number: 659-28-9
EINECS: 211-531-4

Order Now

Specification Test Results

Boiling point 93°C/27mm
Density 1.331
ND20 1.458

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR