1-Iodo-2-(trifluoromethoxy)benzene ≥98%

Supplier: Matrix Scientific

005363-25G 005363-100G
101788-754EA 26.15 USD
101788-754 101788-756
1-Iodo-2-(trifluoromethoxy)benzene ≥98%
1-Iodo-2-(trifluoromethoxy)benzene

Hazard code(s): Irritant

Formula: C₇H₄F₃IO
MW: 288.01 g/mol
Boiling Pt: 74…76 °C (15mm)
MDL Number: MFCD00042410
CAS Number: 175278-00-9

Order Now

Specification Test Results

Boiling point 74-76°C/15m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR